5-Fluoro-4-iodo-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

62512-500MG 62512-1G 62512-5G 62512-25G
77331-682EA 161.71 USD
77331-682 77345-914 77367-818 77389-212
5-Fluoro-4-iodo-2-methoxyphenol ≥97%
5-Fluoro-4-iodo-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆FIO₂
MW: 268.0242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760873
CAS Number: 2385439-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR