(6-Bromo-2,3-difluoro-4-methylphenyl)trimethylsilane ≥95%
Supplier: AOBChem USA
|
Warning
|
62500-500MG
62500-1G
62500-5G
62500-25G
77331-680EA
207.03
USD
77331-680
77355-906
77367-816
77389-210
(6-Bromo-2,3-difluoro-4-methylphenyl)trimethylsilane ≥95%
(6-Bromo-2,3-difluoro-4-methylphenyl)trimethylsilane
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrF₂Si MW: 279.1965 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34760869 CAS Number: 2764729-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...