1,2,3-Trifluoro-4-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

62498-1G 62498-5G 62498-25G
77341-470EA 88.58 USD
77341-470 77367-814 77379-950
1,2,3-Trifluoro-4-(methylsulfonyl)benzene ≥97%
1,2,3-Trifluoro-4-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₅F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD34760867
CAS Number: 2764734-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR