2,3,4-Trifluoro-5-(methylthio)benzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
62491-500MG
62491-1G
62491-5G
62491-25G
77331-678EA
178.19
USD
77331-678
77346-612
77367-812
77389-208
2,3,4-Trifluoro-5-(methylthio)benzoic acid ≥97%
2,3,4-Trifluoro-5-(methylthio)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₅F₃O₂S Storage Temperature: Ambient |
MDL Number:
MFCD34760861 CAS Number: 2764733-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...