3-Chloro-4',5-difluoro-[1,1'-biphenyl]-2-amine ≥95%

Supplier: AOBChem USA

Warning

62486-500MG 62486-1G 62486-5G 62486-25G
77331-674EA 270.89 USD
77331-674 77355-904 77367-808 77389-206
3-Chloro-4',5-difluoro-[1,1'-biphenyl]-2-amine ≥95%
3-Chloro-4',5-difluoro-[1,1'-biphenyl]-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₈ClF₂N
MW: 239.6484 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760857
CAS Number: 1699247-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR