3-Chloro-5-fluoro-4'-methoxy-[1,1'-biphenyl]-2-amine ≥97%

Supplier: AOBChem USA

Warning

62485-500MG 62485-1G 62485-5G 62485-25G
77331-672EA 270.89 USD
77331-672 77355-902 77367-806 77389-204
3-Chloro-5-fluoro-4'-methoxy-[1,1'-biphenyl]-2-amine ≥97%
3-Chloro-5-fluoro-4'-methoxy-[1,1'-biphenyl]-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClFNO
MW: 251.6839 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760856
CAS Number: 2115198-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR