2'-Amino-3'-chloro-5'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

62484-500MG 62484-1G 62484-5G 62484-25G
77331-670EA 271.92 USD
77331-670 77355-900 77367-804 77389-202
2'-Amino-3'-chloro-5'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥95%
2'-Amino-3'-chloro-5'-fluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉ClFNO
MW: 249.6680 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760855
CAS Number: 2764728-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR