3-Chloro-5-fluoro-4'-methyl-[1,1'-biphenyl]-2-amine ≥95%

Supplier: AOBChem USA

Warning

62483-500MG 62483-1G 62483-5G 62483-25G
77331-668EA 270.89 USD
77331-668 77355-898 77367-802 77389-200
3-Chloro-5-fluoro-4'-methyl-[1,1'-biphenyl]-2-amine ≥95%
3-Chloro-5-fluoro-4'-methyl-[1,1'-biphenyl]-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClFN
MW: 235.6845 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760854
CAS Number: 1692434-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR