3-Bromo-4-fluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62478-500MG 62478-1G 62478-5G
77337-196EA 408.91 USD
77337-196 77355-896 77379-946
3-Bromo-4-fluoro-5-methylbenzaldehyde ≥97%
3-Bromo-4-fluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD28531031
CAS Number: 1780141-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR