1-Bromo-2-fluoro-5-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62471-500MG 62471-1G 62471-5G 62471-25G
77331-664EA 136.99 USD
77331-664 77344-556 77367-798 77389-196
1-Bromo-2-fluoro-5-iodo-3-methylbenzene ≥97%
1-Bromo-2-fluoro-5-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
Storage Temperature: Ambient
MDL Number: MFCD28739619
CAS Number: 1805030-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR