(2-Fluoro-5-methoxy-3-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

62467-500MG 62467-1G
77340-412EA 1033.09 USD
77340-412 77355-892
(2-Fluoro-5-methoxy-3-methylphenyl)methanol ≥95%
(2-Fluoro-5-methoxy-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.1808 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110178
CAS Number: 307975-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR