Methyl 2-fluoro-5-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

62465-500MG 62465-1G 62465-5G 62465-25G
77331-660EA 202.91 USD
77331-660 77355-888 77367-792 77389-192
Methyl 2-fluoro-5-methoxy-3-methylbenzoate ≥97%
Methyl 2-fluoro-5-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD16038788
CAS Number: 1260793-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR