2,4-Difluoro-3-methoxy-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62441-500MG 62441-1G 62441-5G 62441-25G
77331-652EA 257.5 USD
77331-652 77355-878 77367-784 77389-186
2,4-Difluoro-3-methoxy-5-(methylthio)benzaldehyde ≥97%
2,4-Difluoro-3-methoxy-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂S
MW: 218.2204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760840
CAS Number: 2807446-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR