2,4-Difluoro-5-formyl-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62439-500MG 62439-1G 62439-5G 62439-25G
77331-650EA 176.13 USD
77331-650 77346-540 77367-782 77389-184
2,4-Difluoro-5-formyl-3-methoxybenzoic acid ≥97%
2,4-Difluoro-5-formyl-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₂O₄
MW: 216.1383 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760839
CAS Number: 2764734-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR