(3-Ethoxy-2,4-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62428-500MG 62428-1G 62428-5G 62428-25G
77331-646EA 93.73 USD
77331-646 77342-482 77367-778 77379-932
(3-Ethoxy-2,4-difluorophenyl)(methyl)sulfane ≥97%
(3-Ethoxy-2,4-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34760836
CAS Number: 2706786-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR