1-(3-Ethoxy-2,4-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
62424-500MG
62424-1G
62424-5G
62424-25G
77331-644EA
91.67
USD
77331-644
77342-450
77367-774
77379-928
1-(3-Ethoxy-2,4-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Ethoxy-2,4-difluorophenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₁₀H₇F₅O₂ MW: 254.1533 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34760833 CAS Number: 2901102-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...