1-(2,4-Difluoro-3-methoxyphenyl)-2,2,2-trifluoroethanol ≥97%

Supplier: AOBChem USA

Warning

62414-500MG 62414-1G 62414-5G 62414-25G
77331-642EA 149.35 USD
77331-642 77345-316 77367-772 77389-182
1-(2,4-Difluoro-3-methoxyphenyl)-2,2,2-trifluoroethanol ≥97%
1-(2,4-Difluoro-3-methoxyphenyl)-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₉H₇F₅O₂
MW: 242.1426 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28061282
CAS Number: 1823985-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR