(3-(Benzyloxy)-2,4-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62407-500MG 62407-1G 62407-5G 62407-25G
77331-640EA 126.69 USD
77331-640 77344-180 77367-770 77379-926
(3-(Benzyloxy)-2,4-difluorophenyl)(methyl)sulfane ≥97%
(3-(Benzyloxy)-2,4-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂OS
MW: 266.3062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760828
CAS Number: 2764733-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR