2-(Benzyloxy)-1,3-difluoro-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

62406-500MG 62406-1G 62406-5G 62406-25G
77331-638EA 145.23 USD
77331-638 77345-142 77367-768 77389-180
2-(Benzyloxy)-1,3-difluoro-4-methylbenzene ≥95%
2-(Benzyloxy)-1,3-difluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂O
MW: 234.2412 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760827
CAS Number: 947279-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR