1-Bromo-3-fluoro-2-isopropoxy-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

62397-500MG 62397-1G 62397-5G
77331-636EA 337.84 USD
77331-636 77355-876 77379-924
1-Bromo-3-fluoro-2-isopropoxy-5-(trifluoromethyl)benzene ≥95%
1-Bromo-3-fluoro-2-isopropoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₄O
MW: 301.0754 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760825
CAS Number: 2901109-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR