2-(Benzyloxy)-1-bromo-3-fluoro-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

62390-500MG 62390-1G 62390-5G
77337-108EA 407.88 USD
77337-108 77355-872 77379-920
2-(Benzyloxy)-1-bromo-3-fluoro-5-(trifluoromethyl)benzene ≥95%
2-(Benzyloxy)-1-bromo-3-fluoro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉BrF₄O
MW: 349.1182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760822
CAS Number: 1610471-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR