1-(Difluoromethoxy)-2-methyl-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

62385-500MG 62385-1G 62385-5G 62385-25G
77331-632EA 116.39 USD
77331-632 77343-434 77367-764 77379-916
1-(Difluoromethoxy)-2-methyl-3-(trifluoromethyl)benzene ≥97%
1-(Difluoromethoxy)-2-methyl-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇F₅O
MW: 226.1432 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18395382
CAS Number: 1261672-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR