1,5-Dibromo-2-chloro-3-methyl-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

62378-500MG 62378-1G 62378-5G 62378-25G
77331-630EA 99.91 USD
77331-630 77342-852 77367-760 77379-912
1,5-Dibromo-2-chloro-3-methyl-4-(trifluoromethyl)benzene ≥95%
1,5-Dibromo-2-chloro-3-methyl-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₄Br₂ClF₃
MW: 352.3735 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760816
CAS Number: 2807467-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR