4'-Fluoro-2-methyl-3-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62366-500MG 62366-1G 62366-5G
77331-622EA 351.23 USD
77331-622 77355-866 77379-908
4'-Fluoro-2-methyl-3-(trifluoromethyl)-1,1'-biphenyl ≥97%
4'-Fluoro-2-methyl-3-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₄
MW: 254.2228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14700407
CAS Number: 1214369-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR