2,4'-Dimethyl-3-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62365-500MG 62365-1G 62365-5G 62365-25G
77331-620EA 251.32 USD
77331-620 77355-864 77367-752 77389-174
2,4'-Dimethyl-3-(trifluoromethyl)-1,1'-biphenyl ≥97%
2,4'-Dimethyl-3-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃
MW: 250.2589 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760809
CAS Number: 2764732-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR