2-Methyl-3-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62364-500MG 62364-1G 62364-5G 62364-25G
77331-618EA 226.6 USD
77331-618 77355-862 77367-750 77389-172
2-Methyl-3-(trifluoromethyl)-1,1'-biphenyl ≥97%
2-Methyl-3-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃
MW: 236.2323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702138
CAS Number: 1214354-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR