Methyl(2-methyl-3-(trifluoromethyl)phenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62361-1G 62361-5G 62361-25G
77340-788EA 60.77 USD
77340-788 77367-748 77379-906
Methyl(2-methyl-3-(trifluoromethyl)phenyl)sulfane ≥97%
Methyl(2-methyl-3-(trifluoromethyl)phenyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₉F₃S
MW: 206.2279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489193
CAS Number: 1822631-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR