1,2-Dichloro-4-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62349-1G 62349-5G 62349-25G
77340-786EA 60.77 USD
77340-786 77367-746 77379-902
1,2-Dichloro-4-fluoro-5-methylbenzene ≥97%
1,2-Dichloro-4-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂F
MW: 179.0190 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143235
CAS Number: 203059-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR