4-Chloro-2-fluoro-5-iodotoluene ≥95%

Supplier: AOBChem USA

Warning

62348-500MG 62348-1G 62348-5G 62348-25G
77331-616EA 116.39 USD
77331-616 77343-496 77367-744 77379-900
4-Chloro-2-fluoro-5-iodotoluene ≥95%
4-Chloro-2-fluoro-5-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
Storage Temperature: Ambient
MDL Number: MFCD00143281
CAS Number: 202982-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR