(3-Acetyl-5-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

62336-500MG 62336-1G 62336-5G
77339-640EA 605.64 USD
77339-640 77355-856 77379-898
(3-Acetyl-5-methylphenyl)boronic acid ≥97%
(3-Acetyl-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD22544494
CAS Number: 1914057-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR