3-Bromo-5-methyl-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62326-500MG 62326-1G 62326-5G 62326-25G
77331-608EA 252.35 USD
77331-608 77355-852 77367-738 77389-166
3-Bromo-5-methyl-2-(methylthio)benzaldehyde ≥97%
3-Bromo-5-methyl-2-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrOS
Storage Temperature: Ambient
MDL Number: MFCD34760802
CAS Number: 2807469-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR