(2,6-Dibromo-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62321-500MG 62321-1G 62321-5G 62321-25G
77331-604EA 100.94 USD
77331-604 77342-958 77367-734 77379-894
(2,6-Dibromo-4-methylphenyl)(methyl)sulfane ≥97%
(2,6-Dibromo-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Br₂S
Storage Temperature: Ambient
MDL Number: MFCD34760799
CAS Number: 2706328-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR