3-Iodo-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62317-500MG 62317-1G 62317-5G 62317-25G
77331-602EA 219.39 USD
77331-602 77355-846 77367-732 77389-162
3-Iodo-5-methylbenzaldehyde ≥97%
3-Iodo-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇IO
Storage Temperature: Ambient
MDL Number: MFCD18397775
CAS Number: 1261585-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR