(3-Bromo-5-methylphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

62314-500MG 62314-1G 62314-5G 62314-25G
77331-598EA 121.54 USD
77331-598 77343-960 77367-728 77379-888
(3-Bromo-5-methylphenyl)(morpholino)methanone ≥97%
(3-Bromo-5-methylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrNO₂
MW: 284.1490 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27728743
CAS Number: 1851578-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR