(3-Bromo-5-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

62313-500MG 62313-1G 62313-25G 62313-5G
77331-596EA 293.55 USD
77331-596 77355-844 77389-160 77379-886
(3-Bromo-5-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%
(3-Bromo-5-methylphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrNO
MW: 268.1496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27728741
CAS Number: 1873196-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR