3-Bromo-N-cyclopropyl-5-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

62312-500MG 62312-1G 62312-25G 62312-5G
77331-594EA 303.85 USD
77331-594 77355-842 77389-158 77379-884
3-Bromo-N-cyclopropyl-5-methylbenzamide ≥97%
3-Bromo-N-cyclopropyl-5-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD30314400
CAS Number: 1567106-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR