3-Bromo-N,5-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

62310-500MG 62310-1G 62310-5G 62310-25G
77331-592EA 161.71 USD
77331-592 77345-912 77367-726 77389-156
3-Bromo-N,5-dimethylbenzamide ≥97%
3-Bromo-N,5-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD31811331
CAS Number: 1567064-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR