2-Chloro-1-ethoxy-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

62299-500MG 62299-1G 62299-5G 62299-25G
77331-590EA 233.81 USD
77331-590 77355-840 77367-724 77389-154
2-Chloro-1-ethoxy-4-isopropoxybenzene ≥97%
2-Chloro-1-ethoxy-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClO₂
MW: 214.6886 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760789
CAS Number: 2807456-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR