2-Chloro-1-ethoxy-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

62297-500MG 62297-1G 62297-5G 62297-25G
77331-588EA 190.55 USD
77331-588 77355-838 77367-722 77389-152
2-Chloro-1-ethoxy-4-methoxybenzene ≥97%
2-Chloro-1-ethoxy-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MW: 186.6354 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044062
CAS Number: 1881296-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR