5-Methyl[2,3']bipyridinyl ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Methyl-2,3'-bipyridine

62295-500MG 62295-1G 62295-5G 62295-25G
77331-586EA 260.59 USD
77331-586 77355-836 77367-720 77389-150
5-Methyl[2,3']bipyridinyl ≥97%
5-Methyl[2,3']bipyridinyl

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂
MW: 170.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037514
CAS Number: 26844-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR