5-Methyl-2,4'-bipyridine ≥97%

Supplier: AOBChem USA

Warning

62294-500MG 62294-1G 62294-5G 62294-25G
77331-584EA 138.02 USD
77331-584 77344-606 77367-718 77389-148
5-Methyl-2,4'-bipyridine ≥97%
5-Methyl-2,4'-bipyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD09037515
CAS Number: 106047-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR