5-Methyl-[2,3'-bipyridine]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

62291-500MG 62291-1G 62291-5G
77335-512EA 356.38 USD
77335-512 77355-834 77379-882
5-Methyl-[2,3'-bipyridine]-3-carbaldehyde ≥97%
5-Methyl-[2,3'-bipyridine]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O
MW: 198.2206 g/mol
Storage Temperature: Ambient
CAS Number: 1785766-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR