2-(4-Bromophenyl)-5-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

62289-500MG 62289-1G 62289-25G 62289-5G
77331-582EA 305.91 USD
77331-582 77355-832 77389-146 77379-880
2-(4-Bromophenyl)-5-methylnicotinaldehyde ≥97%
2-(4-Bromophenyl)-5-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD34760785
CAS Number: 1783998-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR