2-(4-Methoxyphenyl)-5-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

62288-500MG 62288-1G 62288-25G 62288-5G
77331-580EA 305.91 USD
77331-580 77355-830 77389-144 77379-878
2-(4-Methoxyphenyl)-5-methylnicotinaldehyde ≥97%
2-(4-Methoxyphenyl)-5-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.2585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760784
CAS Number: 1781717-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR