2-Ethoxy-3,4-difluoro-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

62280-500MG 62280-1G 62280-5G 62280-25G
77331-572EA 121.54 USD
77331-572 77343-958 77367-712 77379-874
2-Ethoxy-3,4-difluoro-1-methoxybenzene ≥97%
2-Ethoxy-3,4-difluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34760780
CAS Number: 2386768-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR