(5-Chloro-2-fluoro-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

62277-500MG 62277-1G 62277-5G
77331-570EA 326.51 USD
77331-570 77355-824 77379-870
(5-Chloro-2-fluoro-1,3-phenylene)bis(methylsulfane) ≥97%
(5-Chloro-2-fluoro-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₈H₈ClFS₂
MW: 222.7305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760779
CAS Number: 2764730-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR