4'-Chloro-3'-methyl-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

62265-500MG 62265-1G 62265-5G 62265-25G
77331-568EA 227.63 USD
77331-568 77355-822 77367-708 77389-136
4'-Chloro-3'-methyl-[1,1'-biphenyl]-3-carbaldehyde ≥97%
4'-Chloro-3'-methyl-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO
MW: 230.6895 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06858687
CAS Number: 885962-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR