4-Chloro-4'-methoxy-3-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62263-500MG 62263-1G 62263-5G 62263-25G
77331-566EA 184.37 USD
77331-566 77347-172 77367-706 77389-134
4-Chloro-4'-methoxy-3-methyl-1,1'-biphenyl ≥97%
4-Chloro-4'-methoxy-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClO
MW: 232.7054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30547504
CAS Number: 644964-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR