4-Chloro-4'-fluoro-3-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62262-500MG 62262-1G 62262-5G 62262-25G
77331-564EA 208.06 USD
77331-564 77355-820 77367-704 77389-132
4-Chloro-4'-fluoro-3-methyl-1,1'-biphenyl ≥97%
4-Chloro-4'-fluoro-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClF
Storage Temperature: Ambient
MDL Number: MFCD14701455
CAS Number: 1214363-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR