4-Chloro-3,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62261-500MG 62261-1G 62261-5G
77340-324EA 900.22 USD
77340-324 77355-818 77379-868
4-Chloro-3,4'-dimethyl-1,1'-biphenyl ≥97%
4-Chloro-3,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Cl
MW: 216.7060 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760773
CAS Number: 2417151-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR